Skip to Content
Merck
All Photos(1)

Key Documents

740314

Sigma-Aldrich

[(Oxido)phenyl(trifluoromethyl)-λ4-sulfanylidene]dimethylammonium tetrafluoroborate

Synonym(s):

N,N-Dimethyl-S-phenyl-S-(trifluoromethyl)sulfoximinium tetrafluoroborate, N-Methyl-N-[oxidophenyl(trifluoromethyl)-·4-sulfanylidene]methanaminium (1:1) tetrafluoroborate tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11BF7NOS
CAS Number:
Molecular Weight:
325.05
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

85-90 °C

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.C[N+](C)=S(=O)(c1ccccc1)C(F)(F)F

InChI

1S/C9H11F3NOS.BF4/c1-13(2)15(14,9(10,11)12)8-6-4-3-5-7-8;2-1(3,4)5/h3-7H,1-2H3;/q+1;-1

InChI key

JQIUQSKIGNTITL-UHFFFAOYSA-N

Application

Reagent for the preparation of monofluoromethyl esters and ethers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service