Skip to Content
Merck
All Photos(2)

Documents

739030

Sigma-Aldrich

Bis[2-(dicyclohexylphosphino)ethyl]amine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H53NP2
CAS Number:
Molecular Weight:
465.67
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: ligand

mp

64-69 °C

functional group

phosphine

SMILES string

C1CCC(CC1)P(CCNCCP(C2CCCCC2)C3CCCCC3)C4CCCCC4

InChI

1S/C28H53NP2/c1-5-13-25(14-6-1)30(26-15-7-2-8-16-26)23-21-29-22-24-31(27-17-9-3-10-18-27)28-19-11-4-12-20-28/h25-29H,1-24H2

InChI key

OQJIHPKBLHGKJN-UHFFFAOYSA-N

Legal Information

Sold under license from Kanata for research purposes only. WO 2004096735.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service