Skip to Content
Merck
All Photos(2)

Documents

738115

Sigma-Aldrich

1-Fluoro-2,4,6-trimethylpyridinium triflate

95%

Synonym(s):

1-Fluoro-sym-collidinium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11F4NO3S
CAS Number:
Molecular Weight:
289.25
Beilstein:
4777118
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Fluorinations

reagent type: oxidant
reaction type: C-H Activation

mp

162-165 °C (lit.)
164-170 °C

storage temp.

2-8°C

SMILES string

[O-]S(=O)(=O)C(F)(F)F.Cc1cc(C)[n+](F)c(C)c1

InChI

1S/C8H11FN.CHF3O3S/c1-6-4-7(2)10(9)8(3)5-6;2-1(3,4)8(5,6)7/h4-5H,1-3H3;(H,5,6,7)/q+1;/p-1

InChI key

PRIGFEJKMMRJSF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Pd(II)-catalyzed para-selective C-H arylation
  • Fluorination reactions
  • Oxidant in oxidatively-induced aryl-CF3 bond-formation
  • Pd(II)-catalyzed C-H olefination reactions

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service