Skip to Content
Merck
All Photos(2)

Key Documents

731919

Sigma-Aldrich

5-Amino-2,4-dichloropyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3Cl2N3
CAS Number:
Molecular Weight:
163.99
Beilstein:
124417
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
EC Index Number:
664-402-3
NACRES:
NA.22

Assay

97%

form

solid

mp

118-123 °C

storage temp.

2-8°C

SMILES string

Nc1cnc(Cl)nc1Cl

InChI

1S/C4H3Cl2N3/c5-3-2(7)1-8-4(6)9-3/h1H,7H2

InChI key

RINHVELYMZLXIW-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service