Skip to Content
Merck
All Photos(1)

Key Documents

728241

Sigma-Aldrich

(Chloroethynyl)benzene

≥95.0% (HPLC)

Synonym(s):

1-Chloro-2-phenylacetylene, 1-Chloro-2-phenylethyne, Phenylethynyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5Cl
CAS Number:
Molecular Weight:
136.58
Beilstein:
1856338
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (HPLC)

form

liquid

storage temp.

−20°C

SMILES string

ClC#Cc1ccccc1

InChI

1S/C8H5Cl/c9-7-6-8-4-2-1-3-5-8/h1-5H

InChI key

GDWZLADUGAKASM-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service