Skip to Content
Merck
All Photos(1)

Documents

71915

Sigma-Aldrich

Sodium 1-propanethiolate

technical, ≥95% (RT)

Synonym(s):

1-Propanethiol sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2SNa
CAS Number:
Molecular Weight:
98.14
Beilstein:
3556024
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Quality Level

Assay

≥95% (RT)

form

crystals

SMILES string

CCCS[Na]

InChI

1S/C3H8S.Na/c1-2-3-4;/h4H,2-3H2,1H3;/q;+1/p-1

InChI key

XVSFHIIADLZQJP-UHFFFAOYSA-M

Other Notes

Salts of alkanethiols are versatile intermediates for the synthesis of various thiol esters and thio ethers. In contrast to the stenchy gaseous or liquid thiols, the salts are easy to handle solids.; Alkanethiolates are strong nucleophiles and thus efficient reagents for SN2 type dealkylations of hindered esters and lactones; Dealkylation of aryl methyl ethers

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L. Testaferri et al.
Synthesis, 751-751 (1983)
P.A. Bartlett et al.
Tetrahedron Letters, 4459-4459 (1970)
G.I. Feutrill et al.
Tetrahedron Letters, 1327-1327 (1970)
W.R. Vaughan et al.
The Journal of Organic Chemistry, 27, 739-739 (1962)
An improved gas chromatographic method for the detection and quantitation of mercaptans in blood.
H A Mardini et al.
Clinica chimica acta; international journal of clinical chemistry, 113(1), 35-41 (1981-06-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service