Skip to Content
Merck
All Photos(1)

Key Documents

717711

Sigma-Aldrich

3,3′,5,5′-Tetrabromo-1,1′-biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Br4
CAS Number:
Molecular Weight:
469.79
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

mp

185-190 °C

SMILES string

Brc1cc(Br)cc(c1)-c2cc(Br)cc(Br)c2

InChI

1S/C12H6Br4/c13-9-1-7(2-10(14)5-9)8-3-11(15)6-12(16)4-8/h1-6H

InChI key

FXJXZYWFJAXIJX-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Lischka et al.
Nanoscale, 10(25), 12035-12044 (2018-06-16)
Even though the surface-assisted dehalogenative coupling constitutes the most abundant protocol in on-surface synthesis, its full potential will only become visible if selectivity issues with polybrominated precursors are comprehensively understood, opening new venues for both organometallic self-assembly and on-surface polymerization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service