Skip to Content
Merck
All Photos(1)

Documents

713929

Sigma-Aldrich

Dichloro[2-(4,5-dihydro-2-oxazolyl)quinoline]palladium(II)

≥97.0% (AT)

Synonym(s):

Pd(quinox)Cl2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10Cl2N2OPd
CAS Number:
Molecular Weight:
375.55
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (AT)

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
core: palladium

reagent type: ligand

SMILES string

Cl[Pd]Cl.C1CN=C(O1)c2ccc3ccccc3n2

InChI

1S/C12H10N2O.2ClH.Pd/c1-2-4-10-9(3-1)5-6-11(14-10)12-13-7-8-15-12;;;/h1-6H,7-8H2;2*1H;/q;;;+2/p-2

InChI key

DQWIOWASGZSKNG-UHFFFAOYSA-L

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service