Skip to Content
Merck
All Photos(1)

Documents

707457

Sigma-Aldrich

Benzyl chloroformate-(carbonyl-13C)

99 atom % 13C, 97% (CP)

Synonym(s):

Carbobenzoxy chloride-(carbonyl-13C)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C713CH7ClO2
CAS Number:
Molecular Weight:
171.59
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

liquid

bp

103 °C/20 mmHg

density

1.202 g/mL at 25 °C

mass shift

M+1

storage temp.

−20°C

SMILES string

Cl[13C](=O)OCc1ccccc1

InChI

1S/C8H7ClO2/c9-8(10)11-6-7-4-2-1-3-5-7/h1-5H,6H2/i8+1

InChI key

HSDAJNMJOMSNEV-VJJZLTLGSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

197.6 °F

Flash Point(C)

92 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service