Skip to Content
Merck
All Photos(2)

Key Documents

705845

Sigma-Aldrich

4,4′-Bipiperidine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2
CAS Number:
Molecular Weight:
168.28
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

Assay

96%

form

solid

mp

170-172 °C

SMILES string

C1CC(CCN1)C2CCNCC2

InChI

1S/C10H20N2/c1-5-11-6-2-9(1)10-3-7-12-8-4-10/h9-12H,1-8H2

InChI key

PRNRUOJLUPUJDN-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service