Skip to Content
Merck
All Photos(1)

Key Documents

702935

Sigma-Aldrich

2,4,6-Trimethoxyphenylboronic acid neopentyl glycol ester

97%

Synonym(s):

2-(2,4,6-Trimethoxyphenyl)-5,5-dimethyl-1,3,2-dioxaborinane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21BO5
Molecular Weight:
280.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
To inquire about this product 702935, please contact your local Merck office or dealer. Contact Technical Service

Assay

97%

form

powder

mp

105-109 °C

SMILES string

COc1cc(OC)c(B2OCC(C)(C)CO2)c(OC)c1

InChI

1S/C14H21BO5/c1-14(2)8-19-15(20-9-14)13-11(17-4)6-10(16-3)7-12(13)18-5/h6-7H,8-9H2,1-5H3

InChI key

ALIYPFTURKMCNA-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service