Skip to Content
Merck
All Photos(1)

Key Documents

693294

Sigma-Aldrich

1,1,2,2-Tetrafluoroethanesulfonic acid

97%

Synonym(s):

TFESA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CHF2CF2SO3H
CAS Number:
Molecular Weight:
182.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.354

bp

210-211 °C/760 mmHg

density

1.747 g/mL at 25 °C

SMILES string

OS(=O)(=O)C(F)(F)C(F)F

InChI

1S/C2H2F4O3S/c3-1(4)2(5,6)10(7,8)9/h1H,(H,7,8,9)

InChI key

KZWJWYFPLXRYIL-UHFFFAOYSA-N

Application

Alternative to triflic acid.[1]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vsevolod V Rostovtsev et al.
The Journal of organic chemistry, 73(2), 711-714 (2007-12-19)
We have recently developed an improved synthetic route to 1,1,2,2-tetrafluoroethanesulfonic acid (HCF2CF2SO3H, TFESA) and explored the applications of this newly available superacid in catalysis. Low volatility, ease of handling, and a convenient 1H NMR handle make this acid an attractive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service