Skip to Content
Merck
All Photos(1)

Documents

688959

Sigma-Aldrich

(R,R)-Bicyclo[2.2.1]heptane-2,5-dione

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
Beilstein:
9477671
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.5% (GC)
98%

form

powder

optical activity

[α]/D 4.5±1.0°, c = 1 in chloroform

SMILES string

O=C1C[C@H]2C[C@@H]1CC2=O

InChI

1S/C7H8O2/c8-6-2-4-1-5(6)3-7(4)9/h4-5H,1-3H2/t4-,5-/m1/s1

InChI key

RVFBNLMXCXFOET-RFZPGFLSSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chiral diene ligands enable asymmetric transformations, constructing enantioenriched compounds from achiral substrates efficiently.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service