Skip to Content
Merck
All Photos(3)

Key Documents

683906

Sigma-Aldrich

2-Acetamidophenylboronic acid

≥95%

Synonym(s):

(2-Acetylaminophenyl)boronic acid, 2-Carbamoylbenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CONH)C6H4B(OH)2
CAS Number:
Molecular Weight:
178.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

mp

>300 °C

functional group

amide

storage temp.

2-8°C

SMILES string

CC(=O)Nc1ccccc1B(O)O

InChI

1S/C8H10BNO3/c1-6(11)10-8-5-3-2-4-7(8)9(12)13/h2-5,12-13H,1H3,(H,10,11)

InChI key

UMOPBIVXPOETPG-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride.

Application

Reactant for preparation of:
  • Highly substituted acenes via rhodium-catalyzed oxidative coupling with alkynes
  • Anticancer pyrimidines with multiple-kinase inhibitory effect
  • 3,4-ring fused 7-azaindoles and deazapurines as potent JAK2 inhibitors
  • Polycyclic azaindoles as JAK2 inhibitors
  • ROS1 receptor tyrosine kinase inhibitors
  • 2-aminopyrazolopyridines as Polo-like kinase 1 inhibitors

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service