Skip to Content
Merck
All Photos(1)

Key Documents

679143

Sigma-Aldrich

1-Fmoc-3-oxopiperazine

97%

Synonym(s):

1-(9-Fluorenylmethoxycarbonyl)-3-oxopiperazine, 4-Fmoc-2-oxopiperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N2O3
CAS Number:
Molecular Weight:
322.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

124 °C

functional group

Fmoc

SMILES string

O=C1CN(CCN1)C(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C19H18N2O3/c22-18-11-21(10-9-20-18)19(23)24-12-17-15-7-3-1-5-13(15)14-6-2-4-8-16(14)17/h1-8,17H,9-12H2,(H,20,22)

InChI key

YQKHEDWUIMBPKY-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service