Skip to Content
Merck
All Photos(1)

Documents

674826

Sigma-Aldrich

4-(4-Fluorophenyl)-2-(trifluoromethyl)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10F4O2
CAS Number:
Molecular Weight:
250.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

impurities

related aromatic impurities
3.6% ethyl acetate

mp

72-76 °C

SMILES string

OC(=O)C(CCc1ccc(F)cc1)C(F)(F)F

InChI

1S/C11H10F4O2/c12-8-4-1-7(2-5-8)3-6-9(10(16)17)11(13,14)15/h1-2,4-5,9H,3,6H2,(H,16,17)

InChI key

BPFKBGMJHBFEOE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service