669709
2,3-Bis[(2R,5R)-2,5-dimethylphospholano]-N-benzylmaleimide(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
95%
Synonym(s):
3,4-Bis[(2R,5R)-2,5-dimethylphospholanyl]-1-benzyl-1H-pyrrol-2,5-dion(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, catASium® MNBn(R)Rh
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C31H43BF4NO2P2Rh
CAS Number:
Molecular Weight:
713.34
MDL number:
UNSPSC Code:
12352000
Assay
≥95% (31P-NMR)
95%
SMILES string
[Rh+].F[B-](F)(F)F.C1CC=CCCC=C1.C[C@@H]2CC[C@@H](C)P2C3=C(P4[C@H](C)CC[C@H]4C)C(=O)N(Cc5ccccc5)C3=O
General description
sold in collaboration with Solvias AG
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Legal Information
catASium is a registered trademark of Umicore AG & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service