Skip to Content
Merck
All Photos(1)

Key Documents

668524

Sigma-Aldrich

(+)-1,2-Bis[(2R,5R)-2,5-diisopropylphospholano]benzene

kanata purity

Synonym(s):

(R,R)-i-Pr-DUPHOS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44P2
CAS Number:
Molecular Weight:
418.58
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

optical activity

[α]20/D +103°, c = 1 in chloroform

refractive index

n20/D 1.5701

SMILES string

CC(C)[C@H]1CC[C@H](C(C)C)P1c2ccccc2P3[C@H](CC[C@@H]3C(C)C)C(C)C

InChI

1S/C26H44P2/c1-17(2)21-13-14-22(18(3)4)27(21)25-11-9-10-12-26(25)28-23(19(5)6)15-16-24(28)20(7)8/h9-12,17-24H,13-16H2,1-8H3/t21-,22-,23-,24-/m1/s1

InChI key

RBVGOQHQBUPSGX-MOUTVQLLSA-N

General description

Optical rotation devation ± 13

Application

(R,R)-i-Pr-DUPHOS is a chiral phospholane class of ligand that can be used in:
  • The catalytic asymmetric hydrogenation reactions.
  • Rhodium catalyzed enantioselective synthesis of 1,2 diols via asymmetric intramolecular hydrosilylation of α- and β-hydroxyketones.
  • In the enantioselective synthesis of diarylmethanols by arylation of aromatic aldehydes with arylboronic acids in the presence of a rhodium catalyst.

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective hydrogenation of imines using a diverse library of ruthenium dichloride (diphosphine)(diamine) precatalysts
Cobley CJ and Henschke JP
advanced synthesis and catalysis, 345(1-2), 195-201 (2003)
Asymmetric hydrogenation of a 4, 4-diaryl-3-butenoate; a novel approach to sertraline
Boulton LT, et al.
Organic & Biomolecular Chemistry, 1(7), 1094-1096 (2003)
Enantioselective hydrogenation of imines using a diverse library of ruthenium dichloride (diphosphine)(diamine) precatalysts
Cobley CJ and Henschke JP
Advanced Synthesis & Catalysis, 345(1-2), 195-201 (2003)
Asymmetric synthesis of diarylmethanols by Rh (I)-catalyzed arylation of aromatic aldehydes with arylboronic acid
Suzuki K, et al.
Synthesis, 2006(08), 1360-1364 (2006)
Asymmetric intramolecular hydrosilylation of hydroxyketones
Burk MJ and Feaster JE
Tetrahedron Letters, 33(16), 2099-2102 (1992)

Articles

Asymmetric hydrogenation enables scalable synthesis of single-enantiomer compounds with minimal byproducts, ideal for commercial manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service