Skip to Content
Merck
All Photos(1)

Key Documents

663271

Sigma-Aldrich

1,4-Bis(diphenylamino)benzene

97%

Synonym(s):

N,N,N′,N′-Tetraphenyl-benzene-1,4-diamine, Tetra-N-phenyl-p-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H24N2
CAS Number:
Molecular Weight:
412.52
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

solid

mp

201-205 °C

SMILES string

c1ccc(cc1)N(c2ccccc2)c3ccc(cc3)N(c4ccccc4)c5ccccc5

InChI

1S/C30H24N2/c1-5-13-25(14-6-1)31(26-15-7-2-8-16-26)29-21-23-30(24-22-29)32(27-17-9-3-10-18-27)28-19-11-4-12-20-28/h1-24H

InChI key

JPDUPGAVXNALOL-UHFFFAOYSA-N

Application

Used in OLED multilayers[1] and molecular magnets[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strohriegl, P.; et al.
Advanced Mat., 14, 1439-1452 (2002)
Selby et al.
Organic letters, 2(2), 171-174 (2000-05-18)
[structure: see text] Triamino-s-triazine derivatives 3a, 4, and 5 have been prepared, and their cationic states have been analyzed electrochemically. At 298 K, 3a+ has a limited lifetime in CH2Cl2 solution. However, 4+ and 5+ are long-lived under such conditions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service