Skip to Content
Merck
All Photos(1)

Key Documents

662380

Sigma-Aldrich

Lumidot CdS

440, core-type quantum dots, 5 mg/mL in toluene

Synonym(s):

Cadmium monosulfide, Cadmium sulfide yellow

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdS
CAS Number:
Molecular Weight:
144.48
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

Quality Level

form

dispersion

concentration

5 mg/mL in toluene

particle size

2.9-4.0 nm

density

0.865 g/mL at 25 °C

λmax

410-430 nm

fluorescence

λem 430-450 nm FWHM 20 nm

storage temp.

2-8°C

InChI

1S/Cd.S

InChI key

CJOBVZJTOIVNNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Organically stabilized cadmium sulfide quantum dots in toluene

Analysis Note

Particle size was determined by TEM according to the procedure in Yu, W. W., et al., Chem. Mater., 2003, 15, 2854.
Quantum yield was measured following the procedure of Qu, L., Peng, X., J. Am. Chem. Soc., 2002, 124-2049.

Other Notes

Lifetime one year if stored properly

Legal Information

Product of Nanoco Technologies sold under US and worldwide patents and patents pending.
Lumidot is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shancheng Yan et al.
Journal of nanoscience and nanotechnology, 13(6), 4242-4246 (2013-07-19)
A roll-on transfer strategy is developed to enable large-scale and uniform assembly of CdS nanowires on various rigid and flexible substrate materials. In this method, the CdS nanowires were synthesized by the hydrothermal method. The dispersed CdS NWs could be
Janusz Szeremeta et al.
Nanoscale, 5(6), 2388-2393 (2013-02-12)
Wide wavelength range (550-1600 nm) measurements of nonlinear optical properties of ca. 5 nm CdS quantum dots (QDs) colloidal solution were performed using the Z-scan technique with femtosecond laser pulses at 1 kHz. Parameters characterizing nonlinear absorption and nonlinear refraction
C Javaux et al.
Nature nanotechnology, 8(3), 206-212 (2013-02-12)
Applications of semiconductor nanocrystals such as biomarkers and light-emitting optoelectronic devices require that their fluorescence quantum yield be close to 100%. However, such quantum yields have not been obtained yet, in part, because non-radiative Auger recombination in charged nanocrystals could
Yi-Hsin Liu et al.
Journal of the American Chemical Society, 134(45), 18797-18803 (2012-10-26)
Colloidal CdTe quantum wires are reported having ensemble photoluminescence efficiencies as high as 25% under low excitation-power densities. High photoluminescence efficiencies are achieved by formation of a monolayer CdS shell on the CdTe quantum wires. Like other semiconductor nanowires, the
Ou Chen et al.
Nature materials, 12(5), 445-451 (2013-02-05)
High particle uniformity, high photoluminescence quantum yields, narrow and symmetric emission spectral lineshapes and minimal single-dot emission intermittency (known as blinking) have been recognized as universal requirements for the successful use of colloidal quantum dots in nearly all optical applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service