Skip to Content
Merck
All Photos(1)

Key Documents

659215

Sigma-Aldrich

Neodymium(III) iodide

anhydrous, powder, 99.9%

Synonym(s):

Neodymium triiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NdI3
CAS Number:
Molecular Weight:
524.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.9%

form

powder

reaction suitability

reagent type: catalyst
core: neodymium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

SMILES string

I[Nd](I)I

InChI

1S/3HI.Nd/h3*1H;/q;;;+3/p-3

InChI key

DKSXWSAKLYQPQE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service