Skip to Content
Merck
All Photos(1)

Documents

646245

Sigma-Aldrich

Glyceryl tri(oleate-9,10-13C2)

99 atom % 13C, 97% (CP)

Synonym(s):

Triolein-9,10-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C51H104O6
Molecular Weight:
891.39
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

bp

235-240 °C (lit.)

density

0.917 g/mL at 25 °C

mass shift

M+6

storage temp.

2-8°C

SMILES string

CCCCCCCC\[13CH]=[13CH]/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\[13CH]=[13CH]/CCCCCCCC)OC(=O)CCCCCCC\[13CH]=[13CH]/CCCCCCCC

InChI

1S/C57H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-30,54H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27-/i25+1,26+1,27+1,28+1,29+1,30+1

InChI key

PHYFQTYBJUILEZ-VJJJIHCLSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service