Skip to Content
Merck
All Photos(3)

Key Documents

642908

Sigma-Aldrich

2-Methoxy-5-methylbenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.5430 (lit.)

bp

160 °C/15 mmHg (lit.)

density

1.063 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

COc1ccc(C)cc1C#N

InChI

1S/C9H9NO/c1-7-3-4-9(11-2)8(5-7)6-10/h3-5H,1-2H3

InChI key

MYOASWSDRGHGLN-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service