Skip to Content
Merck
All Photos(2)

Key Documents

634263

Sigma-Aldrich

Praseodymium(III,IV) oxide

nanopowder, <200 nm particle size (BET), 99% trace metals basis

Synonym(s):

Hexapraseodymium undecaoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pr6O11
CAS Number:
Molecular Weight:
1021.44
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: praseodymium

particle size

<200 nm (BET)
<50 nm (XRD)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

O=[Pr]=O.O=[Pr]=O.O=[Pr]=O.O=[Pr]=O.O=[Pr]O[Pr]=O

InChI

1S/11O.6Pr

InChI key

AICMLAQRDYRMRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Used in the solid state synthesis of a remarkable ternary, reduced molybdenum oxide, Pr4Mo9O18, whose structure contains previously unknown Mo7, Mo13and Mo19 clusters. The new cluster product is a small band gap semiconductor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service