Skip to Content
Merck
All Photos(1)

Key Documents

634034

Sigma-Aldrich

4-Chloro-2-methylbenzoic acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)CO2H
CAS Number:
Molecular Weight:
170.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 634034, please contact your local Merck office or dealer. Contact Technical Service

Assay

96%

form

solid

mp

167-171 °C (lit.)

functional group

carboxylic acid
chloro

SMILES string

Cc1cc(Cl)ccc1C(O)=O

InChI

1S/C8H7ClO2/c1-5-4-6(9)2-3-7(5)8(10)11/h2-4H,1H3,(H,10,11)

InChI key

XXFKOBGFMUIWDH-UHFFFAOYSA-N

General description

4-Chloro-2-methylbenzoic acid can be synthesized from 4-chlorobenzoic acid via a multi-step reaction process.[1]

Application

4-Chloro-2-methylbenzoic acid can be used in the synthesis of:
  • 4-chloro-2-methylbenzophenone via Freidel Craft′s acylation with benzene[2]
  • 4-chloro-2-methyl-5-(methylsulfonyl)benzoic acid[1]
  • 4-chloro-2-methyl-3-nitrobenzoic acid methyl ester[1]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"A novel synthesis of 1-substituted 2H-isoquinolin-3-ones"
Canepa.SA and Bravo.DR
Journal of Heterocyclic Chemistry, 41(06), 979-982 (2004)
"(2-Methyl-5-(methylsulfonyl) benzoyl) guanidine Na+/H+ antiporter inhibitors"
Baumgarth M, et al.
Journal of Medicinal Chemistry, 40(13), 2017- 2034 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service