Skip to Content
Merck
All Photos(1)

Key Documents

606553

Sigma-Aldrich

Toluene-1-13C

99 atom % 13C

Synonym(s):

Methylbenzene-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC6H8
Molecular Weight:
93.13
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

bp

110 °C (lit.)

mp

-93 °C (lit.)

density

0.874 g/mL at 25 °C

mass shift

M+1

SMILES string

C[13c]1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3/i7+1

InChI key

YXFVVABEGXRONW-CDYZYAPPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service