Skip to Content
Merck
All Photos(1)

Key Documents

595926

Sigma-Aldrich

3-Methyl-2-phenyl-4-pyrazolecarboxylic acid

97%

Synonym(s):

3-Methyl-2-phenylpyrazole-4-carboxylic acid, 5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10N2O2
CAS Number:
Molecular Weight:
202.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

166-170 °C (lit.)

SMILES string

Cc1c(cnn1-c2ccccc2)C(O)=O

InChI

1S/C11H10N2O2/c1-8-10(11(14)15)7-12-13(8)9-5-3-2-4-6-9/h2-7H,1H3,(H,14,15)

InChI key

USSMIQWDLWJQDQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service