Skip to Content
Merck
All Photos(1)

Key Documents

59061

Sigma-Aldrich

Luviquat FC 370

~40% active ingredients in H2O

Synonym(s):

Poly[(3-methyl-1-vinylimidazolium chloride)-co-(1-vinylpyrrolidone)], Polyquaternium D16

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

cationic

concentration

~40% active ingredients in H2O

refractive index

n20/D 1.415

pH

5.0-8.0 (10% in H2O, active ingredients)

density

1.10 g/mL at 20 °C

InChI

1S/C6H9N2.C6H9NO.ClH/c1-3-8-5-4-7(2)6-8;1-2-7-5-3-4-6(7)8;/h3-6H,1H2,2H3;2H,1,3-5H2;1H/q+1;;/p-1

InChI key

ZIQRJGXRRBOCEI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Luviquat FC 370 is also known as polyquaternium-16. It is a cationic copolymer of methylvinylimidazolium chloride and vinylpyrrolidone.

Application

Luviquat FC 370 may be used in cosmetics, stabilizers, lubricants, coatings, bactericides, hair care and skin care products.

Legal Information

Luviquat is a trademark of BASF SE

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Preservatives null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service