Skip to Content
Merck
All Photos(1)

Key Documents

590568

Sigma-Aldrich

Tetrakis(dimethylamido)silane

electronic grade, 99.9999%

Synonym(s):

Octamethylsilanetetramine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]4Si
CAS Number:
Molecular Weight:
204.39
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

electronic grade

Assay

99.9999%

bp

196 °C (lit.)

mp

15 °C (lit.)

density

0.884 g/mL at 25 °C (lit.)

SMILES string

CN(C)[Si](N(C)C)(N(C)C)N(C)C

InChI

1S/C8H24N4Si/c1-9(2)13(10(3)4,11(5)6)12(7)8/h1-8H3

InChI key

SSCVMVQLICADPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service