Skip to Content
Merck
All Photos(1)

Documents

575496

Sigma-Aldrich

3-[2,5-Bis(chloromethyl)-4-methoxyphenoxy]-1-propanesulfonyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3COC6H2(O(CH2)3SO2Cl)(CH2Cl)2
CAS Number:
Molecular Weight:
361.67
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98%

form

solid

mp

97 °C (dec.) (lit.)

SMILES string

COc1cc(CCl)c(OCCCS(Cl)(=O)=O)cc1CCl

InChI

1S/C12H15Cl3O4S/c1-18-11-5-10(8-14)12(6-9(11)7-13)19-3-2-4-20(15,16)17/h5-6H,2-4,7-8H2,1H3

InChI key

JBLWGGLWXFFETE-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna D Peters et al.
Organic & biomolecular chemistry, 16(35), 6479-6490 (2018-08-30)
A bis(cyclam)-capped cholesterol lipid designed to bind C-X-C chemokine receptor type 4 (CXCR4) was synthesised in good overall yield from 4-methoxyphenol through a seven step synthetic route, which also provided a bis(cyclam) intermediate bearing an octaethyleneglycol-primary amine that can be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service