Skip to Content
Merck
All Photos(1)

Documents

57151

Sigma-Aldrich

Indium(III) sulfate

anhydrous, ≥98.0% (T)

Synonym(s):

Diindium trisulfate, Indium sulfate, Indium sulfate (In2 (SO4 )3)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2(SO4)3
CAS Number:
Molecular Weight:
517.82
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98.0% (T)

form

powder

reaction suitability

reagent type: catalyst
core: indium

SMILES string

[In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2In.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6

InChI key

XGCKLPDYTQRDTR-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

  • Multifunctional alginate–sulfonate–silica sphere-shaped adsorbent particles for the recovery of Indium (III) from secondary resources: Discusses the use of indium(III) sulfate in the study of adsorption capacities for indium recovery (J Roosen, S Mullens, K Binnemans, 2017, ACS Publications).
  • Effect of the deposition conditions on the anion resin exchange precipitation of indium (III) hydroxide: This study uses indium(III) sulfate to investigate the precipitation of indium hydroxide via anion resin exchange (N Evsevskaya et al., 2020, ACS Publications).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service