Skip to Content
Merck
All Photos(1)

Documents

570443

Sigma-Aldrich

Disperse Red 13 acrylate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19ClN4O4
CAS Number:
Molecular Weight:
402.83
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98%

form

solid

mp

74-79 °C (lit.)

λmax

202 nm

storage temp.

2-8°C

SMILES string

CCN(CCOC(=O)C=C)c1ccc(cc1)\N=N\c2ccc(cc2Cl)[N+]([O-])=O

InChI

1S/C19H19ClN4O4/c1-3-19(25)28-12-11-23(4-2)15-7-5-14(6-8-15)21-22-18-10-9-16(24(26)27)13-17(18)20/h3,5-10,13H,1,4,11-12H2,2H3/b22-21+

InChI key

MXXVPCCKVUYGHS-QURGRASLSA-N

General description

NLO material

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service