Skip to Content
Merck
All Photos(1)

Key Documents

562432

Sigma-Aldrich

Bis(tetramethylcyclopentadienyl)chromium(II)

97%

Synonym(s):

1,1′,2,2′,3,3′,4,4′-Octamethylchromocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26Cr
CAS Number:
Molecular Weight:
294.40
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: chromium
reagent type: catalyst

mp

106-111 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Cr].C[C]1[CH][C](C)[C](C)[C]1C.C[C]2[CH][C](C)[C](C)[C]2C

InChI

1S/2C9H13.Cr/c2*1-6-5-7(2)9(4)8(6)3;/h2*5H,1-4H3;

InChI key

KVFUREAYKUNDIT-UHFFFAOYSA-N

Application

Investigations of chromium valence by electron energy loss spectroscopy

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service