Skip to Content
Merck
All Photos(1)

Key Documents

549096

Sigma-Aldrich

5-Bromo-2-fluorobenzotrifluoride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)CF3
CAS Number:
Molecular Weight:
243.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.462 (lit.)

bp

79-80 °C/50 mmHg (lit.)

density

1.72 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(Br)cc1C(F)(F)F

InChI

1S/C7H3BrF4/c8-4-1-2-6(9)5(3-4)7(10,11)12/h1-3H

InChI key

AJLIJYGWAXPEOK-UHFFFAOYSA-N

General description

5-Bromo-2-fluorobenzotrifluoride is commonly used in the preparation poly(arylene ether)s (PAEs).

Application

5-Bromo-2-fluorobenzotrifluoride may be used in the preparation of:
  • 3,5-bis(4-fluoro-3-trifluoromethylphenyl)phenol
  • 9,9-bis(2-ethylhexyl)-2,7-bis[4-fluoro-3-trifluoromethylphenyl]-9H-fluorene
  • 3-trifluoromethyl-4-fluoro phenyl boronic acid

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sterically encumbered poly (arylene ether) s containing spiro-annulated substituents: Synthesis and thermal properties.
Huang WY, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 48(24), 5872-5884 (2010)
Extremely high molar mass hyperbranched poly (arylene ether) s from a new semifluorinated AB2 monomer by an unusual AB2+ A2 polymerization approach.
Ghosh A, et al.
Macromolecules, 43(6), 2846-2854 (2010)
Fluorinated poly (arylene ether)s with aliphatic chain appended cardo moiety: Synthesis and gas transport properties.
Ghosh S and Banerjee S.
Journal of Membrane Science, 470, 535-546 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service