Skip to Content
Merck
All Photos(1)

Key Documents

544515

Sigma-Aldrich

5-Bromo-2-methoxybenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(OCH3)SO2Cl
CAS Number:
Molecular Weight:
285.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

115-118 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc(Br)cc1S(Cl)(=O)=O

InChI

1S/C7H6BrClO3S/c1-12-6-3-2-5(8)4-7(6)13(9,10)11/h2-4H,1H3

InChI key

IXSBNNRUQYYMRM-UHFFFAOYSA-N

General description

5-Bromo-2-methoxybenzenesulfonyl chloride is a benzenesulfonyl chloride derivative.

Application

5-Bromo-2-methoxybenzenesulfonyl chloride may be used in the preparation of the following 1-bromo-3-heteroarylbenzene derivatives:
  • 2-(5-bromo-2-methoxyphenyl)benzofuran
  • 2-(5-bromo-2-methoxyphenyl)-1-methylpyrrole
  • 2-(5-bromo-2-methoxyphenyl)benzoxazole

It may also be used to synthesize N-(4-ethylbenzoyl)-2-methoxybenzenesulfonamide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of symmetrical and unsymmetrical 1,3-diheteroarylbenzenes through palladium-catalyzed direct arylation of benzene-1,3-disulfonyl dichloride and 3-bromobenzenesulfonyl chlorides.
Idris I, et al.
Tetrahedron, 71(52), 9617-9625 (2015)
Alina Fomovska et al.
Antimicrobial agents and chemotherapy, 56(5), 2666-2682 (2012-02-23)
Toxoplasma gondii is a protozoan parasite that can damage the human brain and eyes. There are no curative medicines. Herein, we describe our discovery of N-benzoyl-2-hydroxybenzamides as a class of compounds effective in the low nanomolar range against T. gondii

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service