Skip to Content
Merck
All Photos(1)

Key Documents

543020

Sigma-Aldrich

5-Fluorocytidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12FN3O5
CAS Number:
Molecular Weight:
261.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]20/D +55°, c = 1% in methanol

mp

196-200 °C (lit.)

SMILES string

NC1=NC(=O)N(C=C1F)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O

InChI

1S/C9H12FN3O5/c10-3-1-13(9(17)12-7(3)11)8-6(16)5(15)4(2-14)18-8/h1,4-6,8,14-16H,2H2,(H2,11,12,17)/t4-,5-,6-,8-/m1/s1

InChI key

STRZQWQNZQMHQR-UAKXSSHOSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xian-Run Luo et al.
Asian Pacific journal of cancer prevention : APJCP, 13(3), 781-784 (2012-05-29)
The aim of this study was to evaluate the effect of the adenovirus-mediated double suicide gene (CD/TK) for selective killing of gastric cancer cells. Gastric cancer cells SCG7901 and normal gastric epithelial cell lines were infected by adenoviruses Ad-survivin/GFP and
K Randerath et al.
Recent results in cancer research. Fortschritte der Krebsforschung. Progres dans les recherches sur le cancer, 84, 283-297 (1983-01-01)
5-Fluoropyrimidines and 5-azapyrimidines were found in our laboratory to be specific inhibitors of modification reactions taking place at the 5 position of pyrimidines in nucleic acids. Thus, 5-fluorouracil and 5-fluorouridine specifically inhibit the formation of 5-methyluracil, pseudouridine, and 5,6-dihydrouracil in
K A Watanabe et al.
Journal of medicinal chemistry, 24(7), 893-897 (1981-07-01)
5'-O-Glucuronides of anticancer nucleosides, 5-fluorouridine and 5-fluorocytidine, were synthesized by three different methods. The best preparative procedure was the one starting from benzyl 5-O-(methyl 2', 3', 4'-tri-O-acetyl-beta-D-glucopyranosyluronate)-2,3-O-isopropylidene-beta-D-ribof uranoside (15) that was obtained almost quantitatively by condensation of benzyl 2,3-O-isopropylidene-beta-D-ribofuranoside (8)
I I Kaiser et al.
Biochimica et biophysica acta, 825(1), 12-20 (1985-05-24)
RNAs isolated from Escherichia coli B grown in the presence of 5-fluorouracil have high levels of the analog replacing uridine and uridine-derived modified nucleosides. Cytidine has also been shown to be replaced in these RNAs by 5-fluorocytidine, a metabolic product
S Balakrishna Pai et al.
Antiviral chemistry & chemotherapy, 16(3), 183-192 (2005-07-12)
The clinical emergence of lamivudine and adefovir resistance mutations on prolonged therapy further necessitates the development of additional drugs for the treatment of hepatitis B virus (HBV) infections. We have evaluated a number of novel 2'-fluoro-2',3'-unsaturated D- and L-nucleosides for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service