Recommended Products
refractive index
n20/D 1.5400 (lit.)
bp
275-276 °C (lit.)
density
1.223 g/mL at 25 °C (lit.)
SMILES string
CCCc1ccc(cc1)S(Cl)(=O)=O
InChI
1S/C9H11ClO2S/c1-2-3-8-4-6-9(7-5-8)13(10,11)12/h4-7H,2-3H2,1H3
InChI key
LEFGAGRZHLNPLS-UHFFFAOYSA-N
General description
4-N-Propylbenzenesulfonyl chloride is an R-arylsulfonyl chloride.
Application
4-N-Propylbenzenesulfonyl chloride may be used in the synthesis of sodium 4n-propylbenzenesulfonate and N-(2-aminophenyl)arylsulfonamide.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis, characterization and catalytic properties of novel palladium (II) complexes containing aromatic sulfonamides: effective catalysts for the oxidation of benzyl alcohol.
Applied Organometallic Chemistry, 27(1), 52-58 (2013)
Association of hydrotropes in aqueous solution studied by reaction kinetics.
Advanced Synthesis & Catalysis, 344(3/4), 413-420 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service