Skip to Content
Merck
All Photos(2)

Documents

531995

Sigma-Aldrich

2-Chloro-N-benzylacetamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHCOCH2Cl
CAS Number:
Molecular Weight:
183.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

93-96 °C (lit.)

SMILES string

ClCC(=O)NCc1ccccc1

InChI

1S/C9H10ClNO/c10-6-9(12)11-7-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,12)

InChI key

SRAXAXHQMCQHSH-UHFFFAOYSA-N

General description

2-Chloro-N-benzylacetamide can be prepared by reacting benzylamine and chloroacetylchloride.

Application

2-Chloro-N-benzylacetamide may be used in the preparation of:
  • 2-[4-(Aryl substituted) piperazin-1-yl]-N-benzylacetamides by reacting with corresponding arylpiperazines.
  • N-Methylacetamide and N-benzylacetamide by reacting with di-tert-butyl 1,4,7-triazacyclononane-1,4-diacetate.
  • bis[N-Benzyl-2-(quinolin-8-yl­oxy)acetamide] monohydrate by reacting with 8-hydroxyquinoline.
  • N-Benzyl-2-(1H-imidazol-1-yl)acetamide by reacting with imidazole.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation and Characterization of Gallium (III) Complexes with Monoamide Derivatives of 1,4,7-Triazacyclononane-1,4,7-triacetic Acid: A Study of the Dependency of Structure on Reaction pH.
Shetty D, et al.
European Journal of Inorganic Chemistry, 2010(34), 5432-5438 (2010)
Bis [N-benzyl-2-(quinolin-8-yloxy) acetamide] monohydrate.
Wang MS, et al.
Acta Crystallographica Section E, Structure Reports Online, 67(7), o1558-o1558 (2011)
Chiral Variation of a Hybrid Bis (carbene-amido) Ligand System.
Jean-Baptiste dit Dominique F, et al.
Organometallics, 29(13), 2868-2873 (2010)
Synthesis, computational studies and preliminary pharmacological evaluation of 2-[4-(aryl substituted) piperazin-1-yl]-N-benzylacetamides as potential antipsychotics.
Kumar S, et al.
Arabian Journal of Chemistry (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service