Skip to Content
Merck
All Photos(1)

Key Documents

529087

Sigma-Aldrich

2-Chloro-5-iodotoluene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(I)CH3
CAS Number:
Molecular Weight:
252.48
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.624 (lit.)

bp

239 °C (lit.)

density

1.81 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(I)ccc1Cl

InChI

1S/C7H6ClI/c1-5-4-6(9)2-3-7(5)8/h2-4H,1H3

InChI key

MMBDKGFWRIYSRD-UHFFFAOYSA-N

General description

2-Chloro-5-iodotoluene is a halogenated hydrocarbon.

Application

2-Chloro-5-iodotoluene may be used to synthesize:
  • 1-(4-chloro-3-methylphenyl)-1H-pyrrolo[2,3-c]pyridine[1]
  • bromomethyl-2-chloro-5-iodobenzene[2]
  • 3-(4′-chloro-3′-tolyl)thiophene[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frédéric Stauffer et al.
Bioorganic & medicinal chemistry letters, 22(5), 1860-1863 (2012-02-18)
Aromatase inhibition is the new standard of care for estrogen receptor positive breast cancer and has also potential for treatment of other diseases such as endometriosis. Simple and readily available 3-pyridyl arylethers and 1-aryl pyrrolo[2,3-c]pyridines recapitulating the key pharmacophore elements
Synthesis and characterization of all-conjugated graft copolymers comprised of n-type or p-type backbones and poly (3-hexylthiophene) side chains.
Wang J, et al.
Macromolecules, 46(5), 1783-1793 (2013)
Susanna Tchilibon et al.
Journal of medicinal chemistry, 48(6), 1745-1758 (2005-03-18)
A series of ring-constrained (N)-methanocarba-5'-uronamide 2,N(6)-disubstituted adenine nucleosides have been synthesized via Mitsunobu condensation of the nucleobase precursor with a pseudosugar ring containing a 5'-ester functionality. Following appropriate functionalization of the adenine ring, the ester group was converted to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service