Skip to Content
Merck
All Photos(1)

Key Documents

527602

Sigma-Aldrich

(E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester

Synonym(s):

1-[cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)]hexene, 2,2′-[(1Z)-1-Butyl-1,2-ethenediyl]bis[4,4,5,5-tetramethyl-1,3,2-dioxaborolane]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34B2O4
CAS Number:
Molecular Weight:
336.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

refractive index

n20/D 1.455 (lit.)

bp

292 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

[H]\C(B1OC(C)(C)C(C)(C)O1)=C(/CCCC)B2OC(C)(C)C(C)(C)O2

InChI

1S/C18H34B2O4/c1-10-11-12-14(20-23-17(6,7)18(8,9)24-20)13-19-21-15(2,3)16(4,5)22-19/h13H,10-12H2,1-9H3/b14-13-

InChI key

SACOFNGKSPZAFQ-YPKPFQOOSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service