Skip to Content
Merck
All Photos(1)

Documents

521027

Sigma-Aldrich

trans-1-Octen-1-ylboronic acid

≥95%

Synonym(s):

(2E)-2-Octen-1-ylboronic acid, (E)-1-Octen-1-ylboronic acid, (E)-1-Octenylboronic acid, trans-1-Octenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH=CHB(OH)2
CAS Number:
Molecular Weight:
156.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

mp

100-104 °C (lit.)

SMILES string

CCCCCC\C=C\B(O)O

InChI

1S/C8H17BO2/c1-2-3-4-5-6-7-8-9(10)11/h7-8,10-11H,2-6H2,1H3/b8-7+

InChI key

RBTAJLKAPFBZDQ-BQYQJAHWSA-N

Application

Reactant for:
  • Chiral palladacycle-catalyzed asymmetric ring-opening reaction
  • Enantioselective conjugate addition catalyzed by acyltartaric acids
  • Copper-mediated oxidative cross-coupling reactions
  • Diastereoselective domino Heck-Suzuki reactions

Other Notes

Contains varying amounts of anhydride

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service