Skip to Content
Merck
All Photos(2)

Documents

483990

Sigma-Aldrich

Erbium(III) acetylacetonate hydrate

97%

Synonym(s):

2,4-Pentanedione erbium(III) derivative, Er(acac)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Er(C5H7O2)3 · xH2O
CAS Number:
Molecular Weight:
464.58 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder or crystals

reaction suitability

core: erbium
reagent type: catalyst

mp

125 °C (dec.) (lit.)

SMILES string

[H]O[H].CC(=O)\C=C(\C)O[Er](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Er.H2O/c3*1-4(6)3-5(2)7;;/h3*3,6H,1-2H3;;1H2/q;;;+3;/p-3/b3*4-3-;;

InChI key

FDDVCZNWLBKVFL-KJVLTGTBSA-K

Application

The product may be used as one of the constituents used in the preparation of tris(acetylacetonate)mono(bathophenanthroline)erbium(III), [Er(acac)3(bath)]. It may be be used for the preparation of mono dispersed lanthanide oxide nanodisks.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure, theoretical studies and luminescent properties of a ternary erbium (III) complex with acetylacetone and bathophenanthroline ligands.
Martin-Ramos P, et al.
Journal of Luminescence, 162, 41-49 (2015)
Generic synthetic route to monodisperse sub-10 nm lanthanide oxide nanodisks: a modified digestive ripening process.
Jeong J, et al.
Chemistry of Materials, 28(1), 172-179 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service