Skip to Content
Merck
All Photos(1)

Documents

480770

Sigma-Aldrich

Bismuth subgallate hydrate

99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5BiO6 · xH2O
CAS Number:
Molecular Weight:
394.09 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103

Assay

99.9%

mp

223 °C (dec.) (lit.)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service