Skip to Content
Merck
All Photos(3)

Documents

478652

Sigma-Aldrich

4-Biphenylyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4NCO
CAS Number:
Molecular Weight:
195.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

110-112 °C/1 mmHg (lit.)

mp

59-62 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=Nc1ccc(cc1)-c2ccccc2

InChI

1S/C13H9NO/c15-10-14-13-8-6-12(7-9-13)11-4-2-1-3-5-11/h1-9H

InChI key

WIRPZDICFIIBRF-UHFFFAOYSA-N

Related Categories

General description

4-Biphenylyl isocyanate, also known as 4-isocyanatobiphenyl, is an aryl isocyanate. Its enthalpy of vaporization at boiling point has been reported. Its density and refractive index are 1.1266g/cm3 and 1.6060, respectively at 25°C.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 406-406 (2015)
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 605-605 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service