Skip to Content
Merck
All Photos(1)

Key Documents

478261

Sigma-Aldrich

5-Chloro-2-methylphenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)NCO
CAS Number:
Molecular Weight:
167.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.556 (lit.)

bp

115 °C/5 mmHg (lit.)

density

1.226 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccc(Cl)cc1N=C=O

InChI

1S/C8H6ClNO/c1-6-2-3-7(9)4-8(6)10-5-11/h2-4H,1H3

InChI key

XEMUTFNBAICJEO-UHFFFAOYSA-N

General description

5-Chloro-2-methylphenyl isocyanate, also known as 4-chloro-2-isocyanato-1-methylbenzene, is an organic building block containing an isocyanate group. Its Raman spectra, infrared spectra, ab initio and density functional theory (DFT) calculations have been reported.[1]

Application

5-Chloro-2-methylphenyl isocyanate may be used in the synthesis of:
  • N-(5-chloro-2-methyphenyl)-N′-(1-methylethyl)-thiourea[2]
  • N-(5-chloro-2-methyphenyl)-N′-(1-methylethyl)-urea[2]
  • 5-chloro-2-methylphenylcarbamoylated cyclodextrin (CD) chiral stationary phase (CSP)[3]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuzhou Lin et al.
Journal of chromatography. A, 1406, 342-346 (2015-07-04)
In this work, two cyclodextrin (CD) chiral stationary phases (CSPs) have been developed by clicking per-4-chloro-3-methylphenylcarbamoylated mono-6(A)-azido-β-CD (CSP1) and per-5-chloro-2-methylphenylcarbamoylated mono-6(A)-azido-β-CD (CSP2) onto alkynylated silica support. The enantioslectivies of the as-obtained new CSPs have been evaluated using 29 model racemates
Thomas J F Nieland et al.
Journal of lipid research, 48(8), 1832-1845 (2007-05-30)
Treatment of atherosclerotic disease often focuses on reducing plasma LDL-cholesterol or increasing plasma HDL-cholesterol. We examined in vitro the effects on HDL receptor [scavenger receptor class B type I (SR-BI)] activity of three classes of clinical and experimental plasma HDL-cholesterol-elevating
The vibrational spectra, assignments and ab initio/DFT analysis for 3-chloro, 4-chloro and 5-chloro-2-methylphenyl isocyanates.
Doddamani SB, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 67(1), 150-159 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service