Skip to Content
Merck
All Photos(1)

Key Documents

476897

Sigma-Aldrich

Dichloro(diisopropylamino)borane

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NBCl2
CAS Number:
Molecular Weight:
181.90
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

liquid

reaction suitability

reagent type: reductant

bp

61 °C/13 mmHg (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CC(C)N(B(Cl)Cl)C(C)C

InChI

1S/C6H14BCl2N/c1-5(2)10(6(3)4)7(8)9/h5-6H,1-4H3

InChI key

SQJYZIVNKOIKKX-UHFFFAOYSA-N

Application

Reactant for:
  • Cyclizative alkenylboration of carbon-carbon double bonds
  • Cyclizative carboboration of C-C triple bonds
  • Preparation of cationic terminal borylene complexes

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service