Skip to Content
Merck
All Photos(1)

Key Documents

474614

Sigma-Aldrich

Tetrakis[1-[(4-tert-butylphenyl)sulfonyl]-(2S)-pyrrolidinecarboxylate]dirhodium(II)

Synonym(s):

Rh2(S-TBSP)4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H80N4O16Rh2S4
CAS Number:
Molecular Weight:
1447.37
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

optical activity

[α]20/D −187°, c = 0.1 in chloroform

mp

273 °C (dec.) (lit.)

SMILES string

CC(C)(C)c1ccc(cc1)S(=O)(=O)N2CCC[C@H]2C(=O)O[Rh]OC(=O)[C@@H]3CCCN3S(=O)(=O)c4ccc(cc4)C(C)(C)C.CC(C)(C)c5ccc(cc5)S(=O)(=O)N6CCC[C@H]6C(=O)O[Rh]OC(=O)[C@@H]7CCCN7S(=O)(=O)c8ccc(cc8)C(C)(C)C

InChI

1S/4C15H21NO4S.2Rh/c4*1-15(2,3)11-6-8-12(9-7-11)21(19,20)16-10-4-5-13(16)14(17)18;;/h4*6-9,13H,4-5,10H2,1-3H3,(H,17,18);;/q;;;;2*+2/p-4/t4*13-;;/m0000../s1

InChI key

HVKPLPQGLBYKLH-WYIKOJPOSA-J

Application

Catalyst for asymmetric cyclopropanations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davies, H.M.L. Hutcheson, D.K.
Tetrahedron Letters, 34, 7243-7243 (1993)
Davies, H.M.L. et al.
Tetrahedron Letters, 38, 1741-1741 (1997)
Davies, H.M.L. et al.
Tetrahedron Letters, 37, 4133-4133 (1996)
Davies, H.M.L. et al.
Journal of the American Chemical Society, 118, 6897-6897 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service