Skip to Content
Merck
All Photos(1)

Documents

468630

Sigma-Aldrich

4-Fluoro-L-phenylalanine hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH(NH2)CO2H · HCl
CAS Number:
Molecular Weight:
219.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]24/D −8°, c = 1 in H2O

mp

245-251 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Cl.N[C@@H](Cc1ccc(F)cc1)C(O)=O

InChI

1S/C9H10FNO2.ClH/c10-7-3-1-6(2-4-7)5-8(11)9(12)13;/h1-4,8H,5,11H2,(H,12,13);1H/t8-;/m0./s1

InChI key

ZDECBCKSULAIGP-QRPNPIFTSA-N

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service