Skip to Content
Merck
All Photos(1)

Key Documents

460974

Sigma-Aldrich

N,N′-Dibutyl-1,6-hexanediamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH2)3NH(CH2)3CH3]2
CAS Number:
Molecular Weight:
228.42
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.451 (lit.)

bp

131-133 °C/3 mmHg (lit.)

density

0.821 g/mL at 25 °C (lit.)

SMILES string

CCCCNCCCCCCNCCCC

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G L Kennedy et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 22(6), 425-429 (1984-06-01)
Dibutylhexamethylenediamine (DBHMD), a polymer intermediate, is strongly alkaline and is corrosive to eyes and skin. Its inhalation toxicity was defined in rats in both acute and subchronic studies. A 4-hr LC50 of 0.22 mg/litre was obtained for DBHMD of either
Liviu M Mirica et al.
Journal of the American Chemical Society, 128(8), 2654-2665 (2006-02-24)
The activation of dioxygen (O(2)) by Cu(I) complexes is an important process in biological systems and industrial applications. In tyrosinase, a binuclear copper enzyme, a mu-eta(2):eta(2)-peroxodicopper(II) species is accepted generally to be the active oxidant. Reported here is the characterization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service