Skip to Content
Merck
All Photos(1)

Key Documents

454958

Sigma-Aldrich

mono-2-(Methacryloyloxy)ethyl phthalate

contains ~100 ppm monomethyl ether hydroquinone as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[H2C=C(CH3)CO2CH2CH2O2C]C6H4CO2H
CAS Number:
Molecular Weight:
278.26
EC Number:
MDL number:
UNSPSC Code:
12162002

contains

~100 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.523 (lit.)

bp

232 °C (lit.)

density

1.228 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCCOC(=O)c1ccccc1C(O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Lian Chen
Journal of chromatography. A, 1216(34), 6236-6244 (2009-07-28)
Two open-tubular (OT) capillary electrochromatographic (CEC) columns were prepared by chemically bonding ionizable mono-(2-(methacryloyloxy)ethyl) succinate (MES) and phthalate-functionalized (MEP) ligands onto silica hydride-based phases through surface etching, silanization, and hydrosilation reactions, starting with a bare fused-silica tube. An analysis of
Zainuddin et al.
Journal of biomaterials applications, 23(2), 147-168 (2008-07-18)
Converting the surface of poly(2-hydroxyethyl methacrylate) (PHEMA) hydrogel into a cell-adhesive surface has been successfully achieved through a method based on atom transfer radical polymerization (ATRP) grafting. Following activation of the surface hydroxyl groups of PHEMA by bromination, surface-initiated ATRP
G E Schumacher et al.
Dental materials : official publication of the Academy of Dental Materials, 8(4), 278-282 (1992-07-01)
Effective dentin bonding systems based on para-PMDM diadduct of pyromellitic dianhydride and 2-hydroxyethyl methacrylate (HEMA) have been developed (Bowen et al., 1982). Para-PMDM, a solid of limited solubility, is usually applied from an acetone solution to dentin that has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service